Learn More
Sofosbuvir impurity N, MedChemExpress
MedChemExpress Sofosbuvir impurity N, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.
Marca: Medchem Express HY-I0513
Descrizione
Sofosbuvir impurity N, an diastereoisomer of Sofosbuvir, is the impurity of Sofosbuvir. Sofosbuvir (PSI-7977) is an inhibitor of HCV RNA replication, demonstrates potent anti-hepatitis C virus activity.Specifica
Sofosbuvir impurity N | |
White | |
C20H25FN3O9P | |
O=[P@@](N[C@@H](C)C(OC)=O)(OC[C@@H]1[C@@H](O)[C@](F)(C)[C@H](N2C=CC(NC2=O)=O)O1)OC3=CC=CC=C3 | |
501.4 | |
Solid |
1394157-34-6 | |
Research | |
5 mg | |
501.4 | |
Research |