Learn More
Memmert™ Software AtmoCONTROL for SF750plus
AtmoCONTROL Software meets the requirements for the use of electronically stored data sets and electronic signatures.
Marca: Memmert™ SF750plusFDAQ1
Dettagli aggiuntivi : Peso : 0.01200kg
Descrizione
- Software conforming to FDA AtmoCONTROL
- Meets the requirements for the use of electronically stored data sets and electronic signatures as laid down in Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA)
- Base licence for the control of one unit
- Respective IQ/OQ documents available in German and English language (without surcharge)
Specifica
Regulation 21 CFR Part 11 of the US Food and Drug Administration (FDA) | |
SF750plus Sterilizer | |
Software |