Learn More
Merck SPME Fiber Assembly, Carboxen/Polydimethylsiloxane (CAR/PDMS), df 75 μm
For Gases and Low Molecular Weight Compounds (MW 30-225)
Marca: Merck 57318
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Solid phase microextraction (SPME) technique is generally used for GC/MS analysis. It helps in simplifying and speeding up the sample preparation. It also reduces solvent requirement during extraction.Specifica
SPME | |
SPME Fibers & Accessories | |
24 ga |
Manual Holder | |
Fused Silica Fiber | |
3/Pk. |