Learn More
Nickel Electro™ Stainless Steel Bucket with Handle
Sfoglia Tutto Nickel-Electro Prodotti
Click to view available options
Dimensione della confezione:
Pezzo
Descrizione
- 8 Litre Bucket
- Stainless steel construction without graduations
- Bright polished finish for easy cleaning and stainless steel wire handle
- Good chemical resistance and can handle hot liquids
- Stable in use
- Can be autoclaved
Specifica
Specifica
| Product Type | Bucket |
| Materiale | Stainless Steel |
| Autoclavabile | Autoclavable |
| Materiale maniglia | Stainless steel |
| Include | Bright Polished finish |
| Quantità | Ea. |
| Tipo di superficie | Bright Polished |