Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML E2, NVLAP
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Massa:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
Dimensione della confezione:
Pezzo
Descrizione
- Weights in masses ranging from 1 gram to 20 kilograms are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
- Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
- NVLAP Calibration certificate is accredited to ISO 17025 which is recognizable through ILAC
- Weight surfaces meet or exceed OIML R 111 and have a highly polished, mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
Specifica
Specifica
| Certificazione/Conformità | NVLAP Accredited Certificate |
| Materiale | Troemner Alloy 8 |
| Peso (sistema metrico) | 0.300 kg |
| Massa | 200 g |
| Tolleranza | 0.300 mg |
| Include | Protective Case |