Learn More
Ohaus™ Stainless Steel Calibration Weights - OIML F1, NVLAP
Codice prodotto. 16632442 Sfoglia Tutto Ohaus Prodotti
Click to view available options
Massa:
1 mg
2 mg
5 mg
10 mg
20 mg
50 mg
100 mg
200 mg
500 mg
1 g
2 g
5 g
10 g
20 g
50 g
100 g
200 g
500 g
1 kg
2 kg
5 kg
10 kg
20 kg
Dimensione della confezione:
Pezzo
Codice prodotto. 16632442
Marca: Ohaus™ 80780345
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Supplied with a NVLAP Accredited Certificate and Adjusted to Meet or Exceed OIML F1 Tolerances
  • Weights in masses ranging from 1 gram to 1 kilogram are made of Troemner Alloy 8 Stainless Steel which possesses closely controlled density (8.03 g/cm3), extremely low magnetic properties, good stability, and resistance to corrosion
  • Weights are supplied in high quality cases to ensure weights are held securely, kept clean, and dust free
  • NVLAP Calibration Certificate is accredited to ISO 17025 which is recognizable through ILAC
  • Weight surfaces meet or exceed OIML R 111 and have a highly polished mirror-like finish which allows scratches and foreign matters on the weight to be easily seen allowing for stability and longevity
TRUSTED_SUSTAINABILITY
Specifica
Certificazione/Conformità NVLAP Accredited Certificate
Materiale Troemner Alloy 8
Peso (sistema metrico) 0.100 kg
Massa 50 g
Tolleranza 0.300 mg
Include Protective Case