Learn More
Fisherbrand™ Stainless Steel Cylindrical Calibration Weight Set, Class E2 with Certificate
Codice prodotto. 15595119 Sfoglia Tutto Fisherbrand Prodotti
Click to view available options
Peso (sistema metrico):
1 g to 1 kg
1 g to 5 kg
1 to 100 g
200 g
Dimensione della confezione:
1 unità
Codice prodotto. 15595119
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Calibrate weights of lower accuracy classes. Fisherbrand™ Class E2 Individual Stainless Steel Cylindrical Calibration Weights are intended for use in the verification or calibration of class F1 weights and for use with weighing instruments of special accuracy class I.
  • Class E2 stainless steel calibration weight
  • In compliance with OIML R111
  • Delivered with a calibration certificate guaranteeing traceability
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifica
Classificazione E2
Peso (sistema metrico) 1 g to 1 kg
Massa 1 g to 1 kg