Learn More
Edmund Bühler GmbH™ Test Tube Rack
Constructed of Stainless steel. Edmund Bühler GmbH™ Test Tube Rack for use with hinged foot on universal tray or with clamping strip on rack system.
Marca: Edmund Bühler GmbH™ 0052 058
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Stainless steel construction
- Dimensions: 289 x 100mm
- Attach to hinged foot for angle variation (order separately)
- For use with universal platform shakers
Order Info
dimHolds
Specifica
Clamping strips and hinged foot | |
Metal | |
Stainless Steel | |
Non-sterile | |
18 mm | |
18 mm |
Test Tube Rack | |
Tubes (18 mm) | |
44 | |
1 Each | |
289 x 100mm |