Learn More
Karl Hecht™ Stainless Steel Square Wire Baskets
Click to view available options
Size:
16 x 16 x 16 cm
20 x 20 x 20 cm
Dimensione della confezione:
Pezzo
Descrizione
- Wire mesh construction so contents are visible
- Square shape
- Available in a range of sizes
Specifica
Specifica
| Product Type | Wire Basket |
| Descrizione | Wire mesh construction |
| Materiale | Stainless Steel |
| Forma | Square |
| Size | 16 x 16 x 16 cm |