missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Standard Buffer Phosphate, pH 4.002, NIST, Chem-Lab
Click to view available options
Quantità:
1 L
10 L
Dimensione della confezione:
10L
Pezzo
Description
pH = 4.003 at 0°C
pH = 3.99 at 5°C
pH = 3.998 at 10°C
pH = 3.999 at 15°C
pH = 4.002 at 20°C
pH = 4.008 at 25°C
pH = 4.015 at 30°C
pH = 4.035 at 40°C
pH = 4.060 at 50°C
pH = 4.091 at 60°C
pH = 4.126 at 70°C
pH = 4.164 at 80°C
pH = 3.99 at 5°C
pH = 3.998 at 10°C
pH = 3.999 at 15°C
pH = 4.002 at 20°C
pH = 4.008 at 25°C
pH = 4.015 at 30°C
pH = 4.035 at 40°C
pH = 4.060 at 50°C
pH = 4.091 at 60°C
pH = 4.126 at 70°C
pH = 4.164 at 80°C
Specifications
Specifications
| Materiale o nome chimico | Phosphate |
| Densità | 1.00g/mL |
| pH | 4.002 |
| Quantità | 1 L |
| Imballaggio | PE |
| Tracciabilità a NIST | NIST traceable |
| Peso formulazione | 204.23g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction