Learn More
Thermo Scientific™ Manual, Removable-Needle, Gas Tight Syringes for GC Instruments
Codice prodotto. 15224583 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Volume (sistema metrico):
10 μL
25 μL
50 μL
100 μL
250 μL
500 μL
1 mL
5 mL
10 mL
Calibro dell'ago:
22 G
22s G
23 G
25 G
26s G
Stile del punto dell'ago:
90° Blunt
Bevel
Blunt
Side Hole
Dimensione della confezione:
Pezzo
Codice prodotto. 15224583
Marca: Thermo Scientific™ 365M5212
Use Thermo Scientific™ Removable Needle Syringes for accurate and reproducible manual sample introduction into GC instruments, with or without the needle.
Get durability, clarity and accuracy from Thermo Scientific™ Manual Removable Needle Syringes. Used for manual sample injection into gas chromatography (GC) instruments with or without the needle, they provide accuracy and reproducibility that provides confidence in analytical results, time-after-time. Choose from a variety of sizes, and types. These syringes are available in standard industry size with black easy-to-read and solvent-resistant markings.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter.
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
TRUSTED_SUSTAINABILITY
Specifica
Calibro dell'ago 22 G
Lunghezza (sistema metrico) 50 mm
Product Type Gas Tight GC Autosampler Syringe, Removable Needle
Numero dei punti dell'ago 365 RN463
Quantità 1/Ea.
Volume (sistema metrico) 5 mL
Lunghezza (sistema metrico) ago 50 mm
Stile del punto dell'ago Bevel
Da utilizzare con (apparecchiatura) Gas Chromatography Systems
Replacement Needle 365RN463