Learn More
Thermo Scientific™ Sterilin™ Special Petri Dishes
Perform nonstandard applications with Thermo Scientific™ Sterilin™ Special Petri Dishes.
Marca: Thermo Scientific™ 101VRED
Descrizione
Perform nonstandard applications with Thermo Scientific™ Sterilin™ Special Petri Dishes. Minimum order quantities apply.
- Aseptically manufactured
- In vitro use only
Ordering Alerts:
Not available in all geographic regions
Specifica
90mm dishes tested in accordance with EN ISO 24998:2008 standard | |
Red | |
Aseptically Manufactured | |
Piastra di Petri | |
20 | |
16.2 mm |
Piatto | |
Round | |
Non-treated | |
500 | |
3 | |
90 mm |
Suggerimenti di prodotto
I clienti che hanno visualizzato questo articolo hanno visto anche