Learn More
Cole-Parmer™ Stuart™ Stirring Hot Plate and Hot Plate Accessories
Replacement parts and accessories for use with hot plates, stirrers, and/or stirring hot plates.
Marca: Cole-Parmer™ Stuart™ 04805-93
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Replacement parts and accessories for various hot plates, stirrers, and stirring hot plates
Specifica
Temperature Controller | |
180 g | |
Controller-dependent | |
PTFE |
Temperature Probe | |
Controller-dependent | |
Uncoated | |
0.4 lb. |