Learn More
Fisherbrand™ Tappi
Prevents liquid chemical leaks or escape from container.
Marca: Fisherbrand™
Dettagli aggiuntivi : Peso : 0.83250kg
Descrizione
- Small, tapered plug used to seal the openings of test tubes, flasks and other laboratory glassware
- Preferable for applications that require tighter seal or greater degree of chemical resistance
- Prevent contamination of samples by protecting contents of laboratory glassware from air
- Allows experimenters to shake or mix solutions without spilling them
Specifica
Assortiti | |
Per uso di laboratorio | |
Gomma |
Tappo | |
Confezione da 67 |