Learn More
Cole-Parmer™ Bain Marie Storage Container and Snap-Tight, Positive Lock Lid, HDPE
Cole-Parmer™ Bain Marie Storage Container Snap-Tight, Positive-Lock Lid, HDPE is made of FDA compliant HDPE and can be frozen, boiled, or chemically sterilized.
Marca: Cole-Parmer™ 63530-50
Dettagli aggiuntivi : Peso : 0.45000kg
Descrizione
- Durable high-density polyethylene construction
- Bain Marie HDPE containers are sold separately.
- NSF certified.
Specifica
Snap-Tight, Positive-Lock Lid | |
NSF Certified | |
HDPE |