Learn More
Thermo Scientific Chemicals Straight Wall Inconel Crucible;Cap (ml), 45;Outside Dia (mm), 46;Depth (mm), 34
Click to view available options
Capacità (sistema metrico):
5 mL
10 mL
15 mL
20 mL
25 mL
55 mL
75 mL
100 mL
250 mL
500 mL
1000 mL
Diametro (sistema metrico) esterno:
Outside: 101mm
Outside: 127mm
Outside: 21mm
Outside: 27mm
Outside: 33mm
Outside: 45mm
Outside: 46mm
Outside: 47mm
Outside: 51mm
Outside: 59mm
Outside: 82mm
Dimensione della confezione:
Pezzo
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifica
Specifica
| Descrizione | Straight Wall |
| Materiale | Inconel |
| Capacità (sistema metrico) | 25 mL |
| Diametro (sistema metrico) esterno | Outside: 45mm |
| Product Type | Inconel Crucible |