Learn More
Thermo Scientific Chemicals Straight Wall Nickel Crucible;Cap (ml), 35;Outside Dia (mm), 46;Depth (mm), 29
Codice prodotto. p-6685027
Click to view available options
Capacità (sistema metrico):
5 mL
10 mL
15 mL
20 mL
25 mL
35 mL
55 mL
75 mL
100 mL
250 mL
500 mL
Diametro (sistema metrico) esterno:
Outside: 101mm
Outside: 21mm
Outside: 27mm
Outside: 33mm
Outside: 45mm
Outside: 46mm
Outside: 47mm
Outside: 51mm
Outside: 59mm
Outside: 82mm
Dimensione della confezione:
Pezzo
Codice prodotto. 15456135
Marca: Thermo Scientific Chemicals 035889.KT
Richiesta di formato massivo o personalizzato
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Specifica
Descrizione Straight Wall
Materiale Nickel
Capacità (sistema metrico) 5 mL
Diametro (sistema metrico) esterno Outside: 21mm
Altezza (sistema metrico) 29mm
Product Type Nickel Crucible