Learn More
Sulfuric Acid, 10% weight Solution, Chem-Lab
Catalog Number 11949252
Click to view available options
Quantità:
5 L
Imballaggio:
PE
For use in TMAR Ammonium-Phenol and Silicon detection, according to SERES
Materiale o nome chimico Sulfuric acid
CAS 7664-93-9
Concentrazione 107g H2SO4 / 1L H2O
Densità 1.07g/mL
Imballaggio PE
Quantità 5 L
Formula molecolare H2O4S
Numero MDL MFCD00064589
Numero UN UN2796
Sinonimo oil of vitriol, sulphuric acid, dihydrogen sulfate, mattling acid, battery acid, dipping acid, acidum sulfuricum, vitriol brown oil, electrolyte acid
InChI Key QAOWNCQODCNURD-UHFFFAOYSA-N
SMILES OS(O)(=O)=O
IUPAC Name sulfuric acid
Molecular Weight (g/mol) 98.07
PubChem CID 1118
ChEBI CHEBI:26836
Peso formulazione 98.08g/mol
Percent Purity 10% (weight)