Learn More
Thermo Scientific™ Super Clean™ Gas Cartridge Filter Baseplates and Accessories
Select from a range of quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters.
Marca:  Thermo Scientific™ 60180-815
Dettagli aggiuntivi : Peso : 0.50000kg
Codice prodotto. 10544305
  • 715.26€ / Pezzo
Spedizione stimata: 31-10-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Use quality baseplates/manifolds for your Thermo Scientific™ Super Clean™ Cartridge Filters. A range of baseplate options allows configuration to your individual requirements. Needle valves ensure that the gas line is not contaminated during cartridge change. The baseplates have a one-time installation procedure. Super Clean Cartridge Filters are not included.
Additional Product Information
  • Use with Super Clean cartridge filters
  • Filters not included
Specifica
Specifica
2x Combi Filter
For Gas: Air and Fuel Gas
Dual Base