Learn More
OHAUS™ Support Stand
Sfoglia Tutto Ohaus Prodotti
Click to view available options
Diametro (sistema metrico) asta:
16 mm
17 mm
18 mm
19 mm
20 mm
21 mm
22 mm
Altezza (sistema metrico):
267 mm
305 mm
Dimensione della confezione:
Pezzo
Descrizione
- Corrosion-resistant lab-frames, rods and connectors are available in premium stainless steel, offering high chemical resistance and durability. Aluminum offers reliable strength at a lighter weight.
- Simplify lab set up! Thirteen lab-frame kit options are available with four designed to fit in fume hoods. Build your own support system choosing from a wide selection of rods, bases and connectors.
- Lab lifts offer convenient adjustable lifting platforms with exceptional strength and durability for holding small lab equipment such as glassware, hot plates and water baths at different heights.
Specifica
Specifica
| Materiale | Stainless Steel |
| Altezza (sistema metrico) | 305 mm |
| Lunghezza (sistema imperiale) | 12 in. |
| Lunghezza (sistema metrico) | 305 mm |
| Lunghezza (sistema metrico) asta | 1016 mm |
| Larghezza (sistema imperiale) | 16.50 in. |
| Larghezza (sistema metrico) | 419 mm |
| Diametro (sistema metrico) asta | 20 mm |
| Product Type | Support Stand with Rod |