Learn More
Thermo Scientific™ SWAP-IT™ GC-MS Interface System Accessories
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
Marca:  Thermo Scientific™ 290SG002
Dettagli aggiuntivi : Peso : 0.10000kg
Codice prodotto. 10351555
  • 88.34€ / Pezzo
Spedizione stimata: 20-08-2024
per visualizzare lo stock disponibile
Aggiungi al carrello
Descrizione
Descrizione
Find the right nut and ferrule to use with the powerful Thermo Scientific™ SWAP-IT™ GC-MS Interface system.
  • Specially deactivated , robust stainless steel transfer capillary provides an interface between the analytical column and the MS detector
  • Small ID of transfer capillary ensures exposure to air and water is reduced to a minimum during disconnection of the analytical column
  • The system works without the requirement for additional He-purge/make-up gas flow
  • Finger-tight connections make exchanging the analytical column quick and easy without the need for tools
  • Includes finger-tight connections for S/SL injector to provide complete finger-tight solution
Specifica
Specifica
SWAP-Tight Nut
SWAP-Tight Nut
1/Ea.
SWAP-IT GC-MS Interface System