Learn More
Köttermann Tall Safety Storage Cabinet with Doors
Click to view available options
Door Style:
Folding, Manual-close
Hinged, Auto-close
N. di ripiani:
3
4
6
None
Include:
1 Drip Tray
4 Internal Drawers
5 Internal Drawers
Dimensione della confezione:
Pezzo
Descrizione
- Different versions that meet your needs
- Additional storage space
- Top safety for flammable hazardous substances (Type 90)
Specifica
Specifica
| Dimensioni (P x L x A) | 615 x 1200 x 1955 mm |
| Door Style | Hinged, Auto-close |
| Include | 5 Internal Drawers |
| N. di sportelli | 2 |
| N. di ripiani | None |