Learn More
Unity Lab Services™ Tech Direct Service Plan for Thermo Scientific Shakers
Click to view available options
Dimensione della confezione:
Pezzo
Descrizione
- Continue service support post factory warranty with enhanced remote support and annual preventative maintenance
- 3 business day response time (may vary by location) for corrective maintenance
- 15% discount off billable rates for engineer labor, and factory-certified replacement parts for on-site corrective maintenance visit
- One scheduled Preventative Maintenance visit, including a preventative maintenance kit (where applicable)
- Priority unlimited access to OEM-trained Technical support experts who can diagnose, investigate, and remotely resolve issues or schedule on-site visits using the latest digital remote support tools
- Access to on-line service knowledge base
- Corrective Maintenance Repair Guarantee for the duration of product warranty or 1 year whichever is longer, as defined in the contract terms and conditions
- Instrument Software/ Firmware updates included as needed to support a preventative maintenance visit
- Upgrades to new versions of software are not included
- 10% discount on additional services including additional preventative maintenance visits, calibration, and qualification services
Specifica
Specifica
| Da utilizzare con (apparecchiatura) | Shakers |
| Da utilizzare con (applicazione) | Support Post-Factory Warranty |
| Include | 3 Day Response Time (Within Specified Location and Criteria), Priority Technical Support, and One Preventative Maintenance Visit |
| Product Type | Tech Direct Service Plan |
| Services Not Included | Upgrades to New Software Versions |
| Warranty Response Time | 3 Days (Within Specified Location and Criteria) |
| Services Included | Priority Technical Support, and One Preventative Maintenance Visit |