Learn More
Thermo Scientific™ Teledyne CETAC Autosamplers
Increase sample throughput with advanced Teledyne CETAC Autosamplers, compatible with Thermo Scientific™ high-performance trace elemental analyzers.
Marca: Thermo Scientific™ BRE0003260
Descrizione
Increase sample throughput in your laboratory with Teledyne CETAC Autosamplers. This selection of advanced autosamplers, compatible with Thermo Scientific™ high performance trace elemental analyzers, enables operators to take on other tasks, while intelligent, automated sample introduction accelerates workflows and eliminates manual errors.
Teledyne CETAC ASX-280 AutosamplerAccommodates up to 180 sample capacity for low-moderate sample throughput, compatible with:
- Thermo Scientific™ iCE™ 3000 Series AAS systems
- Thermo Scientific™ iCAP™ 7000 Series ICP-OES systems
- Thermo Scientific™ iCAP™ PRO ICP-OES systems
- Thermo Scientific™ iCAP™ Q/Qnova Series ICP-MS systems
- Thermo Scientific™ Element™ Series HR-ICP-MS systems
Accommodates up to 360 samples for moderate-high sample throughput, compatible with:
- iCE 3000 Series AAS systems
- iCAP 7000 Series ICP-OES systems
- iCAP PRO ICP-OES systems
- iCAP Q/Qnova Series ICP-MS systems
- Element Series HR-ICP-MS systems
Enables the unattended analysis of up to 720 samples, for laboratories looking to push the boundaries of unattended analysis, compatible with:
- iCAP 7000 Series ICP-OES systems
- iCAP PRO ICP-OES systems
- iCAP Q/Qnova Series ICP-MS systems
Incorporates automated liquid stirring to ensure sample homogeneity, for samples containing particulates in suspension such as wear metals applications, compatible with:
- iCAP 7000 Series ICP-OES systems
- iCAP PRO ICP-OES systems
Specifica
Autosampler | |
Teledyne CETAC ASX-560 Autosampler for Aqueous Solutions | |
iCE 3000 Series AA, iCAP Series ICP-OES, iCAP Q/Qnova Series ICP-MS, Element HR-ICP-MS |