Learn More
Labconco™ Temperature Sensor for FreeZone™
This Temperature Sensor for FreeZone™ is an RTD sensor for monitoring product sample temperature.
Marca: Labconco™ 7515300
Dettagli aggiuntivi : Peso : 0.45000kg
Descrizione
- Replacement temperature sensor for all Labconco™ FreeZone™ Freeze Dryers
Specifica
Sensor | |
RTD | |
Labconco™ FreeZone™ Freeze Dryers |