missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone-16,16,17-d3, Honeywell Fluka™
missing translation for 'orderingAttributeHoverText'
Quantità:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support
Specifications
Specifications
CAS | 77546-39-5 |
Imballaggio | Glass Bottle |
Quantità | 10 mg |
Grado | Analytical Standard |
Da utilizzare con (applicazione) | Steroids and Hormones |
Forma fisica | Solid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction