Learn More
Texwipe™ Kit convalida pulizia TOC
Sfoglia Tutto Texwipe Prodotti
Click to view available options
Product Type:
Kit di convalida TOC
Validation Kit, Long Handle Swab
Da utilizzare con (applicazione):
Campionamento di 12 aree di produzione
Campionamento di 72 aree di produzione
Sampling 12 Production Areas
Dimensione della confezione:
18 sets
Pezzo
pezzi 18
Descrizione
Includes
Vials, swabs, and labels for sampling 12 or 72 production areas, and container for sample transport.
Designed to simplify sampling for cleaning validation. Kits compatible for use with Sievers Model 800/900, Anatel ANATOC™, Teledyne Tekmar, OI Analytical Model 1010 and Shimadzu TOC-V Series™ TOC Analyzers.
Vials
- Certified Low TOC level (less-than10Glc mug/L, less-than10ppb) ensures consistency in vial background contribution
- Prewashed vials have one-piece cap and septa design for easy sampling and analysis
- Polyethylene overcap protects septum surface from contamination until time of analysis
- Sloped shoulder design minimizes headspace for reduced background TOC level
- Endorsed for use by TOC instrumentation manufacturerbullets1 (end)
- Certified Low TOC level (less-than50Glc mug/L, less-than50ppb) ensures consistency in swab background contribution
- Double layered, double knit polyester head entraps contaminants in knit structure during sampling
- Allows maximum contaminant collection and release into diluent to provide for excellent recovery rates
- Notched, break-away handle allows head to be placed into vial with minimal handling and contamination
- 100% polypropylene handle ensures no additional contaminants are introduced and provides excellent chemical resistance
Certifications
Certificate of Analysis included on each box.
Specifica
Specifica
| Product Type | Kit di convalida TOC |
| Da utilizzare con (applicazione) | Campionamento di 12 aree di produzione |
| Include | 12 fiale trasparenti da 40 ml, setti integrati, 24 tamponi per campionamento Low TOC Alpha™, 12 etichette bianche per fiale |
| Quantità | 18/contenitore |