missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TGFBR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA532631
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 30 min at room temperature. A suggested positive control is placenta tissue.
A total of three members of the TGF beta family, TGFbeta1, TGFbeta2 and TGFbeta3, have been identified in mammals. Each is synthesized as a latent precursor that is subsequently cleaved forming the 112 amino acid growth factor which becomes active upon dimerization. TGFbetas mediate their activity by high affinity binding to the type II receptor 70kDa transmembrane protein with a cytoplasmic serine-threonine kinase domain.Specifica
TGFBR1 | |
Polyclonal | |
TGFBR1 | |
AAT5, ACVRLK4, ALK-5, ALK5, LDS1A, LDS2A, MSSE, SKR4, TGFR-1, TGF-beta receptor type I, TGF-beta receptor type-1, TGF-beta type I receptor, activin A receptor type II-like kinase, 53kD, activin A receptor type II-like kinase, 53kDa, activin A receptor type II-like protein kinase of 53kD, activin receptor-like kinase 5, ser | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
29591, 7046, 21812 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
Unconjugated | |
P80204, P36897, Q64729 | |
TGFBR1 | |
Synthetic peptide corresponding to internal region of human TGF beta R1. | |
500 μL | |
Primary | |
Bovine, Human, Mouse, Rat | |
Antibody | |
IgG |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto