Learn More
Thermo Scientific™ Thermal Mixers and Blocks
Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Product Type:
Block
Mixer with Block
Capacità:
20 x 0.2mL, 12 x 1.5mL Microtubes
20 x 0.5mL, 12 x 1.5mL Microtubes
20 x 2mL, 12 x 1.5mL Microtubes
24 x 1.5mL Microtubes
24 x 2.0mL Microtubes
96-well Microplates, 0.2mL Tube Strips
Dimensione della confezione:
Pezzo
Descrizione
Gain variable speed and temperature control with the Thermo Scientific™ Thermal Mixer, which comes with a choice of blocks for microtubes and microplates.
- Heating and cooling capability to cover a wide range of applications
- Compact size with quiet operation
- Convenient packages that include a Thermal Mixer with a Block
- Heating and cooling capability to cover a wide range of applications
- Compact size with quiet operation
- Convenient packages that include a Thermal Mixer with a Block
Warranty
2 anni
Specifica
Specifica
| Capacità | 20 x 2mL, 12 x 1.5mL Microtubes |
| Product Type | Block |
| Certificazione/Conformità | Certificate of Analysis (COA), Certificate of Conformance (COC) |
| N. di posizioni | 20 x 2 mL (Block), 12 x 1.5 mL (Microtubes) |