Learn More
Texwipe™ ThermaSeal™ 60 Wipers
Sfoglia Tutto Texwipe Prodotti
Click to view available options
Dimensioni (L x P):
10 x 10 cm
Dimensione della confezione:
pezzi 150
Descrizione
- Offer good absorbency, wiping efficiency and abrasion resistance while maintaining chemical compatibility with standard solvents and alcohols
- Wiping and cleaning surfaces, equipment and parts
- Applying and removing lubricants, adhesives, residues and other solutions including disinfectants
- Cleaning with solvents such as isopropyl alcohol (IPA), ethanol, acetone, and degreasers
- Lining trays for holding, protecting, drying and storing of parts, equipment and devices
- Appropriate for use with temperatures less than 400°F (205°C)
Specifica
Specifica
| Materiale | Polyester |
| Dimensioni (L x P) | 10 x 10 cm |
| Quantità confezione | 2 Bags x 75 Wipes, 10 Bags/Cs. |