Learn More
Unity Lab Services™ Thermo Scientific™ Centrifugation Validation Service with Cycle Testing
Stay compliant with our Industry-Exclusive, No-Charge Requalification Guarantee included with each Thermo Scientific Centrifugation Validation Service with Cycle Testing for Benchtop, Micro, Bioprocessing, Clinical, and Standard floor model centrifuges: Thermo Scientific, Sorvall
Marca:  Unity Lab Services™
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 15523679
Questo articolo non è attualmente disponibile o è stato dismesso.
La preghiamo di consultare la pagina del prodotto per individuare le possibili alternative.
Visualizzare prodotti alternativi »
Descrizione
Descrizione
  • Utility, Facility and Environment Verifies that the installation site satisfactorily meets manufacturer-specified environmental requirements.
  • Specifica
    Specifica
    Thermo Scientific Centrifugation including Benchtop, Micro, Bioprocessing, Clinical, and Standard floormodel centrifuges: Thermo Scientific, Sorvall
    Industry-Exclusive, No-Charge Requalification Guarantee. Unity Lab Services is the only OEM service provider to offer a guarantee and promise to requalify instruments and equipment at no charge if a key component fails while under the original factory warranty or a qualifying instrument service plan.
    Travel to major metropolitan areas. Does not include Installation. Instrument must be installed, powered-up and set temperature point for 24 hours before on-site service
    Validation (IQ/OQ) includes a standardized protocol procedure, IQ: Verify Installation and environmental conditions, OQ: Verify operation to design specification, perform min/max speed, brief/extended time and temperature testing (for refrigerated models). Cycle Testing: Point-In-Time test exercise of pre-defined duration that surpasses singular calibration and routine speed, time, and temperature monitoring activities.
    Contact your local sales representative for a list of standard and extended coverage areas.
    Suggerimenti di prodotto
    Suggerimenti di prodotto
    Videos
    SDS
    Documenti
    Documenti
    Certificati
    Vedi i prodotti accessori