Learn More
Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels
Codice prodotto. p-4530725 Sfoglia Tutto Thermo Scientific Prodotti
Click to view available options
Capacità (sistema metrico):
5 L
10 L
20 L
32 L
Dimensione della confezione:
Pezzo
Codice prodotto. 10480394
Marca: Thermo Scientific™ TY509X1
Store and dispense small amounts of liquid nitrogen with our economical and durable liquid nitrogen transfer vessels.
Store and dispense small amounts of liquid nitrogen with our economical and durable Thermo Scientific™ Thermo Series Liquid Nitrogen Transfer Vessels. The Thermo Series includes six models with capacities from 5 to 32 L. A full range of handling and dispensing accessories as well as transport carts are also available.
Features:
  • Designed to bring liquid nitrogen to other cryovessels
  • Lightweight aluminum dewars assure low-static evaporation rates
  • Optional withdrawal device, 12mL dipper and wheeled accessory cart
  • Compact Thermo 5 and 10 dewars feature a convenient pail-style handle for pouring and for use in applications where only small quantities of liquid nitrogen are needed
  • Thermo 10, 20, and 30 can be fitted with self-pressurized withdrawal accessory devices to easily dispense LN2 without pouring
Note:
Stated static evaporation rates are based on new container performance with no lid openings and no product loads. Actual working performance may vary with individual applications.
TRUSTED_SUSTAINABILITY
Specifica
Tipo LN2 Transfer Vessel
Capacità (sistema metrico) 5 L
Altezza (sistema imperiale) esterno 18.2 in.
Altezza (sistema metrico) esterno 46.2 cm
Diametro (sistema imperiale) 8.8 in.
Diametro (sistema imperiale) interno collo 2.2 in.
Diametro (sistema metrico) 22.4 cm
Diameter (Metric) Inside Neck 5.6 cm
Peso di spedizione (sistema imperiale) 12 lb.
Peso di spedizione (sistema metrico) 5.4 kg
Peso (sistema imperiale) 8 lb.
Peso (sistema metrico) 3.6 kg