Learn More
SCAT™ Thread Adapter, Type 81
Sfoglia Tutto SCAT Europe Prodotti
Click to view available options
Dimensione chiusura:
GL45, B83
Dimensione della confezione:
Pezzo
Descrizione
SCAT™ thread adapters are ideal for connecting safety caps, safety waste caps, funnels, canisters or other disposal and supply units.
- Made of polypropylene (PP) material
- Available in a wide range of thread sizes
- Suitable for all SCAT™ caps, funnels or canisters
Specifica
Specifica
| Dimensione chiusura | GL45, B83 |
| Colore chiusura | Gray |
| Materiale rivestimento interno | Polypropylene |
| Product Type | Thread Adapter |