Descrizione
- Universal potentiometric titrator with analogue and IDS digital sensor technology for the determination of pH and ORP values
- Exceptional high visibility colour graphic display for viewing even at extreme angles
- Clear graphic representation of titration curves
- Pre-installed standard methods for potentiometric and KF titration
- USB connectivity
- Ideally suited for water analysis
Specifica
50 user methods | |
3 | |
USB export or pdf/CSV format | |
Online display | |
English | |
TitroLine 7800 | |
0 to 14 | |
0.004 pH units | |
153mm x 296mm x 450mm | |
11.6 in. |
1 | |
1xLAN, 2xUSB-A, 1xUSB-B, 2xRS232 | |
Color VGA TFT display with 320x240 pixels | |
One analogue and one digital (IDS) measuring input | |
Polypropylene Housing | |
1200 mV +\- | |
0.2mV | |
5/10/20/50 mL | |
6.0 in. |