Learn More
WTW™ Kit di analisi per azoto totale
Obtain precision results using the CombiCheck Total Nitrogen NTotal cell ready-made test kit for the determination of total nitrogen in waste water, drinking water, and surface water.
Marca: WTW™ 251996
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
- Variety of instrumentation for testing nitrogen in water
- For waste water, drinking water, surface water
- Nitrospectral Method
- Photometric Test Methods
- CombiCheck Total Nitrogen NTotal cell test kit
- Safe determinations of COD and NTotal in water
Specifica
Chromosulfuric acid oxidation | |
For Photoflex™ photometers | |
5 to 150 mg/L | |
Photometric |
For waste water, drinking water, surface water | |
30 | |
25 Tests | |
Combicheck COD Chemical oxygen demand O2 cell test kit |