Learn More
enQuireBio™ TranslationBlocker Human HIF-1 alpha siRNA
Blocks transcription and translation of HIF-1 alpha from Human.
Marca: enQuireBio™ QX12-2nmol
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_001521 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
3091 | |
2 nmol | |
Human |
Human mda-mb-435 Breast Cancer Cells. | |
HIF1A | |
siRNA | |
HIF-1-alpha, HIF1-ALPHA, HIF1, HIF-1alpha, PASD8, MOP1, HIF-1A, bHLHe78, Hypoxia-inducible factor 1-alpha | |
Lyophilized |