missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ TranslationBlocker Human Notch-1 siRNA
Blocks transcription and translation of Notch-1 from Human.
Marca: enQuireBio™ QX37-10nmol
Questo articolo non è restituibile.
Consulta la politica di reso
Specifica
NP_060087 | |
Store Lyophilized siRNA duplex at 4°C. Reconsituted product may be stored at -20°C for up to 1 month. | |
4851 | |
10 nmol | |
Human |
Ca Ski Human Carcinoma Cells fro Cervix with Integrated HPV-16 genome. | |
NOTCH1 | |
siRNA | |
AOVD1, TAN1, AOS5, hN1, Neurogenic locus notch homolog protein 1 | |
Lyophilized |