Learn More
BTX™ Triple Electroporation Tweezertrode
Click to view available options
Diametro (sistema metrico):
3 mm
5 mm
Dimensione della confezione:
Pezzo
Descrizione
Triple Tweezertrodes have:
- Three electrode contacts with adjustable position and polarities
- Improves electrical field distribution for in utero electroporation applications
Compatability:
- ECM830
- ECM2001+
- Gemini X2
- Connect to BTX generators using the Tweezertrode Connection cables (45-0204)
Applications:
- In vivo drug or gene delivery
- Ex vivo drug or gene delivery
- In utero drug or gene delivery
Tweezertrodes may be cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide.
Specifica
Specifica
| Autoclavabile | Not Autoclavable |
| Tipo | Tweezers |
| Monouso | Reusable |
| Da utilizzare con (apparecchiatura) | ECM 830, ECM 2001, Gemini X2 |
| Sterilità | Non-sterile |
| Metodo di sterilizzazione | Cleaned with a mild detergent and sterilized with 70% ethanol or ethylene oxide. |
| Diametro (sistema imperiale) | 0.2 in. |
| Diametro (sistema metrico) | 5 mm |