Learn More
TrkB Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA588482
338.68 EUR valido fino al 2025-03-29
Usa il codice promo "24111" per avere il tuo prezzo promozionale.
Allerta:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Descrizione
Immunogen sequence: SRIWCSDPSP GIVAFPRLEP NSVDPENITE IFIANQKRLE IINEDDVEAY VGLRNLTIVD SGLKFVAHKA FLKNSNLQHI NFTRNKLTSL SRKHFRHLDL SELILVGNPF TCSCDIMWIK TLQEAKSSPD TQDLYCLNES SKNIPLANLQ IPNCGLPSAN LAAPNLTVEE GKSITLSCSV AGDPVPNMYW DVGNLVSKHM NETSHTQGSL RITNISSDDS GKQISCVAEN LVGEDQDSVN LTVHFAPTIT FLESPTSDHH WCIPFTVKGN PKPALQWFYN GAILNESKYI CTKIHVTNHT EYHGCLQLDN PTHMNNGDYT LIAKNEYGKD EKQISAHFMG WPGIDDGANP NYPDVIYEDY GTAANDIGDT TNRSNEIPST DVTDKTGREH LSV; Positive Samples: K-562, Mouse thymus; Cellular Location: Cell membrane, Endosome membrane, Single-pass type I membrane protein
NTRK2 (TRKB) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system. NTRK2 is the receptor for brain-derived neurotrophic factor (BDNF). Together, NTRK2 and BDNF regulate both short-term synaptic functions and long-term potentiation of brain synapses. TrkB is involved in the development and maintenance of the nervous system. Moreover, Trk B is a tyrosine kinase gene highly related to Trk A. Trk B expression is confined to tissues within the central and peripheral nervous systems. The brain-derived neurotrophic factor (BDNF) and NT-3, but not NGF, can induce rapid phosphorylation on tyrosine of Trk B gp145, one of the receptors encoded by Trk B, although BDNF elicits a response at least two orders of magnitude greater than NT-3. TrkB is a membrane-bound receptor that, upon neurotrophin binding, phosphorylates itself and members of the MAPK pathway. Signaling through TrkB leads to cell differentiation. Mutations in the TrkB gene have been associated with obesity and mood disorders. Alternate transcriptional splice variants encoding different isoforms have been found for the TrkB gene.
Specifica
TrkB | |
Polyclonal | |
Unconjugated | |
Ntrk2 | |
AI848316, BDNF/NT-3 growth factors receptor, BDNF-tropomyosine receptor kinase B, C030027L06Rik, EC 2.7.10.1, GP145 TrkB, GP145TrkB, GP145-TrkB, GP145-TrkB/GP95-TrkB, GP95 TrkB, GP95-TrkB, hyperphagia, kinase TrkB, neural receptor protein-tyrosine kinase (trkB), neurotrophic receptor tyrosine kinase 2, neurotrophic tyrosine kinase receptor type 2, neurotrophic tyrosine kinase, receptor, type 2, neurotrophic tyrosine receptor kinase type 2, NTRK 2, Ntrk2, OTTHUMP00000021573, OTTHUMP00000021574, OTTHUMP00000021576, OTTHUMP00000021577, RATTRKB1, Tkrb, Trk B, TRKB, trk-B, trkB protein, trkB tyrosine kinase, TRKB1, Tropomyosin-related kinase B, tyrosine kinase receptor B | |
Rabbit | |
Affinity Chromatography | |
RUO | |
18212, 4915 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1.7 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal, pH 7.3 | |
P15209, Q16620 | |
Ntrk2 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 41-433 of human TrkB (NP_0010070981) | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.