Learn More
JULABO™ Tube Fitting Adapters
Tube Fitting Adapters are available in various styles to address laboratory needs.
Marca: JULABO™ 8890108
Dettagli aggiuntivi : Peso : 0.27000kg
Descrizione
- Adapter fitting styles include:
- M24x1.5 male to 12 mm barbed fittings
- G 1 in. male to G 3/4 in. female
- G 1 1/2 in. male to G 3/4 in. female
- G 1 in. male to G 3/4 in. male
- G 1 1/2 in. male to G 3/4 in. male
Specifica
G 1 1/2 in. Male to G 3/4 in. Male | |
Fitting Adapter, Fluid Tubing |