Learn More
JULABO™ Tube Fitting Adapters
Tube Fitting Adapters are available in various styles to address laboratory needs.
Marca:  JULABO™ 8890108
Dettagli aggiuntivi : Peso : 0.27000kg
Codice prodotto. 17178498
  • 49.00€ / Pezzo
Spedizione stimata: 27-11-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
  • Adapter fitting styles include:
    • M24x1.5 male to 12 mm barbed fittings
    • G 1 in. male to G 3/4 in. female
    • G 1 1/2 in. male to G 3/4 in. female
    • G 1 in. male to G 3/4 in. male
    • G 1 1/2 in. male to G 3/4 in. male
Specifica
Specifica
G 1 1/2 in. Male to G 3/4 in. Male
Fitting Adapter, Fluid Tubing