Learn More
WTW™ Turb™ Turbidity Standard
AMCO Clear™ standards are polymer calibration standards for Turb™ meters and are much more accurate and stable without drifting than Formazine
Marca: WTW™ 600546
Dettagli aggiuntivi : Peso : 0.25000kg
Descrizione
AMCO Clear™ standards are polymer calibration standards for Turb™ meters and are much more accurate and stable without drifting than Formazine. They are traceable to Formazine according to N.I.S.T and are lot-certified. The long-term stability without particle change in size and shape and the high manufacturing tolerance of only ±1% helps achieve the highest results accuracy. The standards are adapted to the respective device optics in their particle composition. The natural drifting of Formazine and the much higher manufacturing tolerance with Formazine grants them clear advantages. This makes AMCO Clear™ standards especially suitable for applications in the lower measuring range such as drinking water.Specifica
Turb 555 WL Laboratory Turbidimeter | |
Turbidity standard | |
10,000 NTU |