Learn More
Lenz Laborglasintrumente Two-Neck Round-Bottom Flasks, Side Neck Parallel
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Marca: Lenz Laborglasintrumente 03204158
Dettagli aggiuntivi : Peso : 0.10000kg
Descrizione
- Distillation flasks
- With parallel side neck
- With conical ground joints
- With inscription label
- Material: DURAN™
Specifica
500 mL | |
Duran Glass | |
Side Neck NS 14/23 | |
105 mm |