missing translation for 'onlineSavingsMsg'
Learn More
Learn More
U-2 OS (human osterogenic sarcoma) Whole cell lysate, Denatured; Abnova
Marca: Abnova L022V1.200ug
Questo articolo non è restituibile.
Consulta la politica di reso
Descrizione
Western Blotting
Specifica
Western Blot | |
Whole cell lysate (denatured) | |
Cell lysate was prepared by homogenization in ice-cold modified RIPA Lysis Buffer with cocktail of protease inhibitors (Sigma). Cell debris was removed by centrifugation. Protein concentration was determined with Bradford assay (Bio-Rad protein assay, Microplate Standard assay). The lysate was mixed in 5X Sample Buffer to become final 1X Sample Buffer as Storage Buffer. The lysate was heated at 95°C for 5 min, and cooled rapidly. | |
200 μg | |
Human | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Bone | |
Modified RIPA Lysis Buffer: 50 mM Tris-HCl pH 7.4, 150 mM NaCl, 1mM EDTA, 1% Triton X-100, 0.1% SDS, 1% Sodium deoxycholate, 1mM PMSF. | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
In ready-to-use 1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue). | |
3 mg/mL |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto