Learn More
Thermo Scientific Chemicals Ultra thin-bore sample tubes, 1mm ID, <3.98mm OD
For use with MKII Magnetic Susceptibility Balances
Marca:  Thermo Scientific™ 036152.KJ
Dettagli aggiuntivi : Peso : 0.01000kg
Codice prodotto. 15476155
  • 312.00€ / pezzi 5
Spedizione stimata: 27-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • I.D.: 1mm
Specifica
Specifica
5 Pack
Ultra Thin-bore Sample Tubes