Learn More
BICASA Undercounter Drawer Cabinet on Casters
Codice prodotto. 30057562 Sfoglia Tutto BICASA Prodotti
Click to view available options
Colore:
Anthracite
Blue
Light Gray
Red
Dimensioni (P x L x A):
600 x (500 + 20) x 630 mm
600 x (500 + 20) x 760 mm
900 x (500 + 20) x 760 mm
Dimensione della confezione:
Pezzo
Codice prodotto. 30057562
Marca: BICASA TN.DW.0600.760A
Solo null disponibili
Aggiungi al carrello
Aggiungi al carrello
Undercounter Drawer Cabinet on Casters with fire-retardant particleboard body and drawers with soft closing and single lock.
  • Undercounter cabinets on casters
  • Metal frame base with 4 swivel casters (Polyamide case and thermoplastic rubber treads), 2 with lockable with brakes - Height mm 100
  • Central locking system as standard
  • Anti-tilt safety system to open only one drawer at a time
  • Body width mm 500 + mm 20 of ergonomic anodized and painted aluminum handle
Specifica
Product Type Mobile Storage Cabinet
Colore Anthracite
Tipo di armadietto Underbench cabinet
Dimensioni (P x L x A) 600 x (500 + 20) x 760 mm
N. di sportelli 1
N. di cassetti 4
Larghezza (sistema imperiale) 23.6 in.
Larghezza (sistema metrico) 600 mm
Con ruote Yes
Materiale Melamine