Learn More
Unity Lab Services™ Unity Basic Plan Autoclave Group 3
Maximize technology investment by predicting and preventing problems before they occur.
Marca: Unity Lab Services™ ULS_BASIC_ATC-3
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
Basic Support Plan Features:
- Priority status for technical support inquiries
- One (1) Scheduled Preventative Maintenance visit*
- 25% discount on parts required for service calls
- 10% discount on labor and travel required for service calls
- Software updates and notifications
*Parts required for PM visit are not included but 25% discount applies
Specifica
Autoclave Group 3 | |
Basic Plan |