Learn More
Beckman Coulter™ Universal Vial Rubber Caps
Used in combination with Beckman Coulter™ Universal Plastic Vials (mfr. no. A62251). Beckman Coulter™ Universal Vial Rubber Caps are blue, pre-cut, single-use caps that can be used with vials accommodating either run buffer or sample. Pack of 100.
Marca:  Beckman Coulter™ A62250
Encompass
Codice prodotto. 10008099
  • 154.98€ / Pezzo
Spedizione stimata: 19-11-2024
per visualizzare lo stock disponibile
Esplora altre offerte speciali
Aggiungi al carrello
Questo articolo non è restituibile. Consulta la politica di reso
Descrizione
Descrizione
  • Vial caps are one time use only and are not intended for re-use
  • Pre-cut rubber caps allow easy needle access
  • Universal Vials are made of polymethylpentene, have been tested for chemical compatibility, and are for use only PA 800 Enhanced, PA 800 Plus, CESI 8000 Plus, or P/ACE MDQ Plus instruments
Specifica
Specifica
Blue
Yes
100
Rubber
Single-use
Plug Cap