Learn More
Invitrogen™ Ureaplasma parvum Monoclonal Antibody (C46G)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA517010
Descrizione
MA5-17010 has been used in ELISA and IF applications and shows reactivity with the multi-banded antigen (MB) of several Ureaplasma serotypes. MA5-07010 does not cross react with Mycoplasma hominis and Mycoplasma genitallium. MA5-17010 reacts with serovars 1,2, 3, 4, 5, 6, 7, 8, 9, 10, 11, 12 & 14.
Specifica
| Ureaplasma parvum | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% sodium azide; pH 7.2 | |
| Mouse | |
| Protein A | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| ELISA, Western Blot, Immunocytochemistry | |
| C46G | |
| Unconjugated | |
| U. parvum | |
| Reactive with the multi-banded antigen (MB) of several Ureaplasma serotypes. | |
| 1 mL | |
| Primary | |
| 4°C | |
| Liquid |
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.