Learn More
Uvex™ Visor 9707
Click to view available options
Frame Color: Black
Descrizione
- Impact resistant polystyrene
- Built-in head protection
- Field of vision CA is clear
Specifica
Specifica
Lens Coating | Anti-fog |
Lens Tint | Clear |
Product Type | Faceshield |
Colore telaio | Black |
Modello | 9707 |
Materiale | Polycarbonate |
Monouso | Reusable |
Lens Material | Polycarbonate |
Materiale del telaio | Plastic |
UV Protection | UV380 |